Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174465.2 | NP_776890.1 | 5 - 6 | 576 | 77 | |
GenBank mRNA | AF221669.1 | - | 5 - 6 | 576 | 77 | |
BC102089.1 | - | 5 - 6 | 581 | 77 | ||
BC150010.1 | - | 5 - 6 | 605 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |