Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174466.2 | NP_776891.1 | 1 | 649 | 62 | |
GenBank mRNA | AY026859.1 | - | 1 | 649 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |