Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174469.2 | NP_776894.1 | 3 - 4 | 1180 | 77 | |
GenBank mRNA | BC116083.1 | - | 3 - 4 | 1102 | 77 | |
X06295.1 | - | 1180 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons |