Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCKDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174506.1 | NP_776931.1 | 2 - 3 | 421 | 70 | |
GenBank mRNA | BC109669.1 | - | 2 - 3 | 315 | 70 | |
J03759.1 | - | 2 - 3 | 421 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |