Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174519.2 | NP_776944.1 | 1 - 2 | 445 | 58 | |
GenBank mRNA | AB030082.1 | - | 1 - 2 | 445 | 58 | |
AB035210.1 | - | 1 - 2 | 321 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |