Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174524.2 | NP_776949.1 | 1 - 2 | 46 | 99 | |
GenBank mRNA | M60329.1 | - | 1 - 2 | 46 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |