Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174536.2 | NP_776961.1 | 1 - 2 | 78 | 98 | |
GenBank mRNA | AF005940.1 | - | 1 - 2 | 78 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |