Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174541.2 | NP_776966.1 | 6 - 7 | 561 | 95 | |
GenBank mRNA | X12361.1 | - | 561 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |