Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174565.3 | NP_776990.1 | 6 - 7 | 692 | 98 | |
GenBank mRNA | BC102401.1 | - | 6 - 7 | 606 | 98 | |
DT883895.1 | - | 6 - 7 | 704 | 98 | ||
M22539.1 | - | 6 - 7 | 607 | 98 | ||
X14724.1 | - | 465 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |