Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174584.2 | NP_777009.1 | 5 - 6 | 422 | 157 | |
GenBank mRNA | X67154.1 | - | 422 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |