Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174638.3 | NP_777063.2 | 6 - 7 | 1125 | 66 | |
GenBank mRNA | AH003250.2 | - | 6 - 7 | 1123 | 66 | |
D00185.1 | - | 1125 | 66 |
Gene Symbol: LOC787647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | D00361.1 | - | 1127 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |