Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174638.3 | NP_777063.2 | 7 - 8 | 1207 | 65 | |
GenBank mRNA | AH003250.2 | - | 7 - 8 | 1205 | 65 | |
D00185.1 | - | 1207 | 65 |
Gene Symbol: LOC787647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | D00361.1 | - | 1209 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |