Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC24A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174655.2 | NP_777080.1 | 3 - 4 | 2133 | 57 | |
GenBank mRNA | X66481.1 | - | 2133 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |