Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174657.2 | NP_777082.1 | 7 - 8 | 988 | 84 | |
GenBank mRNA | BC149996.1 | - | 7 - 8 | 1075 | 84 | |
X05340.1 | - | 988 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |