Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174672.2 | NP_777097.1 | 2 - 3 | 236 | 61 | |
GenBank mRNA | BC149316.1 | - | 2 - 3 | 333 | 61 | |
BC149338.1 | - | 2 - 3 | 333 | 61 | ||
X66035.1 | - | 236 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |