Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174711.2 | NP_777136.1 | 6 - 7 | 1070 | 134 | |
GenBank mRNA | BC109558.1 | - | 6 - 7 | 1098 | 134 | |
M76543.1 | - | 6 - 7 | 1070 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |