Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174807.1 | NP_777232.1 | 2 - 3 | 109 | 88 | |
GenBank mRNA | M22466.1 | - | 2 - 3 | 109 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |