Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174808.1 | NP_777233.1 | 5 - 6 | 757 | 67 | |
GenBank mRNA | BC118304.1 | - | 5 - 6 | 708 | 67 | |
M58607.1 | - | 5 - 6 | 757 | 67 | ||
M63009.1 | - | 5 - 6 | 777 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |