Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174810.2 | NP_777235.1 | 3 - 4 | 284 | 65 | |
GenBank mRNA | BC102616.1 | - | 3 - 4 | 284 | 65 | |
BT021530.1 | - | 3 - 4 | 297 | 65 | ||
J03244.1 | - | 3 - 4 | 284 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |