Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175050.1 | NP_778220.1 | 7 - 8 | 846 | 109 | |
GenBank mRNA | D26147.1 | - | 7 - 8 | 846 | 109 | |
D26148.1 | - | 7 - 8 | 846 | 109 | ||
D26149.1 | - | 2 - 3 | 210 | 109 | ||
M82978.1 | - | 2 - 3 | 239 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |