Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175050.1 | NP_778220.1 | 9 - 10 | 1104 | 141 | |
GenBank mRNA | D26148.1 | - | 9 - 10 | 1104 | 141 | |
D26149.1 | - | 5 - 6 | 723 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |