Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175797.2 | NP_786991.1 | 3 - 4 | 294 | 134 | |
GenBank mRNA | AF182001.1 | - | 3 - 4 | 294 | 134 | |
BC102109.1 | - | 3 - 4 | 316 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |