Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175814.3 | NP_787008.1 | 1 | 107 | 68 | |
GenBank mRNA | BC102062.1 | - | 1 | 108 | 68 | |
BT025477.1 | - | 2 - 3 | 77 | 68 | ||
S74803.1 | - | 1 | 92 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |