Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175828.2 | NP_787022.1 | 2 - 3 | 288 | 59 | |
GenBank mRNA | BC109777.1 | - | 2 - 3 | 305 | 59 | |
X63216.1 | - | 288 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |