Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175832.2 | NP_787026.1 | 1 - 2 | 331 | 87 | |
GenBank mRNA | AJ237996.1 | - | 1 - 2 | 331 | 87 | |
BC123463.1 | - | 1 - 2 | 309 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |