Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176622.1 | NP_788795.1 | 8 - 9 | 988 | 126 | |
GenBank mRNA | AF020514.1 | - | 8 - 9 | 988 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |