Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176646.3 | NP_788819.1 | 4 - 5 | 1089 | 61 | |
GenBank mRNA | AB026444.1 | - | 4 - 5 | 1090 | 61 | |
BC109553.1 | - | 4 - 5 | 1107 | 61 | ||
BT030731.1 | - | 4 - 5 | 1092 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |