Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176657.1 | NP_788830.1 | 3 - 4 | 439 | 50 | |
GenBank mRNA | AF139106.1 | - | 3 - 4 | 439 | 50 | |
AF141017.1 | - | 3 - 4 | 439 | 50 | ||
AF221522.1 | - | 3 - 4 | 371 | 50 | ||
BC102159.1 | - | 3 - 4 | 477 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |