Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176657.1 | NP_788830.1 | 5 - 6 | 981 | 127 | |
GenBank mRNA | AF139106.1 | - | 5 - 6 | 981 | 127 | |
AF221522.1 | - | 5 - 6 | 913 | 127 | ||
BC102159.1 | - | 5 - 6 | 1019 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |