Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177506.2 | NP_803472.1 | 1 - 2 | 204 | 87 | |
GenBank mRNA | BC120315.1 | - | 1 - 2 | 123 | 87 | |
M80626.1 | - | 1 - 2 | 204 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |