Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180998.2 | NP_851341.1 | 11 - 12 | 1485 | 84 | |
GenBank mRNA | AB046664.1 | - | 11 - 12 | 1346 | 84 | |
BC116051.1 | - | 11 - 12 | 1364 | 84 | ||
EU812318.1 | - | 11 - 12 | 1354 | 84 | ||
FJ589071.1 | - | 11 - 12 | 1352 | 84 | ||
GQ351344.1 | - | 10 - 11 | 1292 | 84 | ||
L08604.1 | - | 11 - 12 | 1485 | 84 | ||
M63502.1 | - | 12 - 13 | 1387 | 84 | ||
X54801.1 | - | 1267 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |