Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180998.2 | NP_851341.1 | 14 - 15 | 1853 | 85 | |
GenBank mRNA | AB046664.1 | - | 14 - 15 | 1714 | 85 | |
BC116051.1 | - | 14 - 15 | 1732 | 85 | ||
EU812318.1 | - | 14 - 15 | 1722 | 85 | ||
FJ589071.1 | - | 14 - 15 | 1720 | 85 | ||
GQ351344.1 | - | 13 - 14 | 1660 | 85 | ||
L08604.1 | - | 14 - 15 | 1853 | 85 | ||
L19981.1 | - | 14 - 15 | 1753 | 85 | ||
M63502.1 | - | 15 - 16 | 1755 | 85 | ||
X17066.1 | - | 655 | 85 | |||
X54801.1 | - | 1635 | 85 | |||
X57084.1 | - | 1746 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |