Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180998.2 | NP_851341.1 | 1 - 2 | 182 | 108 | |
GenBank mRNA | BC116051.1 | - | 1 - 2 | 61 | 108 | |
FJ589071.1 | - | 1 - 2 | 49 | 108 | ||
L08604.1 | - | 1 - 2 | 182 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |