Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181006.2 | NP_851349.1 | 1 - 2 | 167 | 83 | |
GenBank mRNA | BC114913.1 | - | 1 - 2 | 203 | 83 | |
U88551.1 | - | 1 - 2 | 167 | 83 | ||
X55027.1 | - | 175 | 83 | |||
X55489.1 | - | 160 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |