Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181018.2 | NP_851361.1 | 11 - 12 | 1752 | 87 | |
GenBank mRNA | AF001261.1 | - | 11 - 12 | 1752 | 87 | |
AF001262.1 | - | 11 - 12 | 1748 | 87 | ||
AF001263.1 | - | 11 - 12 | 1883 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |