Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181018.2 | NP_851361.1 | 7 - 8 | 1029 | 141 | |
GenBank mRNA | AF001261.1 | - | 7 - 8 | 1029 | 141 | |
AF001262.1 | - | 7 - 8 | 1025 | 141 | ||
AF001263.1 | - | 7 - 8 | 1160 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |