Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181030.2 | NP_851373.1 | 4 - 5 | 347 | 66 | |
GenBank mRNA | AB051841.1 | - | 4 - 5 | 347 | 66 | |
AF282985.1 | - | 4 - 5 | 347 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |