Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181031.2 | NP_851374.1 | 3 - 4 | 610 | 91 | |
GenBank mRNA | BC147979.1 | - | 3 - 4 | 699 | 91 | |
BT021727.1 | - | 3 - 4 | 489 | 91 | ||
M16968.1 | - | 3 - 4 | 588 | 91 | ||
X52554.1 | - | 610 | 91 | |||
X72858.1 | - | 648 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |