Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203361.1 | NP_976237.1 | 5 - 6 | 520 | 78 | |
GenBank mRNA | AY195839.1 | - | 5 - 6 | 520 | 78 | |
BC104567.1 | - | 5 - 6 | 577 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |