Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKDP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205775.1 | NP_991344.1 | 3 - 4 | 250 | 89 | |
GenBank mRNA | AF241779.1 | - | 3 - 4 | 250 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |