Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205817.1 | NP_991386.1 | 1 - 2 | 60 | 70 | |
GenBank mRNA | BC102359.1 | - | 1 - 2 | 62 | 70 | |
X59418.1 | - | 60 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |