Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205817.1 | NP_991386.1 | 3 - 4 | 333 | 63 | |
GenBank mRNA | BC102359.1 | - | 3 - 4 | 335 | 63 | |
BT025401.1 | - | 3 - 4 | 287 | 63 | ||
X59418.1 | - | 333 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |