Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205817.1 | NP_991386.1 | 9 - 10 | 914 | 75 | |
GenBank mRNA | BC102359.1 | - | 9 - 10 | 916 | 75 | |
BT025401.1 | - | 9 - 10 | 868 | 75 | ||
X59418.1 | - | 914 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |