Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001147.1 | NP_001001147.1 | 2 - 3 | 234 | 149 | |
GenBank mRNA | AY428555.1 | - | 2 - 3 | 234 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |