Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001601.1 | NP_001001601.1 | 7 - 8 | 1273 | 75 | |
GenBank mRNA | AY363224.1 | - | 7 - 8 | 1273 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |