Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002884.1 | NP_001002884.1 | 1 - 2 | 108 | 79 | |
GenBank mRNA | AY156928.1 | - | 1 - 2 | 108 | 79 | |
BC149263.1 | - | 1 - 2 | 174 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |