Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012519.1 | NP_001012537.1 | 9 - 10 | 936 | 95 | |
GenBank mRNA | BC110178.1 | - | 9 - 10 | 993 | 95 | |
BT021058.1 | - | 9 - 10 | 936 | 95 | ||
M31463.1 | - | 8 - 9 | 852 | 95 | ||
S54973.1 | - | 9 - 10 | 883 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |