Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPLP0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012682.1 | NP_001012700.1 | 6 - 7 | 715 | 71 | |
GenBank mRNA | AB098748.1 | - | 2 - 3 | 371 | 71 | |
AB098788.1 | - | 2 - 3 | 371 | 71 | ||
AB099074.1 | - | 2 - 3 | 230 | 71 | ||
AF013214.1 | - | 4 - 5 | 608 | 71 | ||
BC102074.1 | - | 6 - 7 | 729 | 71 | ||
BC151695.1 | - | 6 - 7 | 727 | 71 | ||
BT021080.1 | - | 6 - 7 | 715 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Endogenous Controls Recommended Assay Bos taurus Probe spans exons |