Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014388.1 | NP_001014410.1 | 5 - 6 | 619 | 108 | |
GenBank mRNA | AB098755.1 | - | 1 - 2 | 205 | 108 | |
BC104562.1 | - | 5 - 6 | 628 | 108 | ||
BT021036.1 | - | 5 - 6 | 619 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |