Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014867.1 | NP_001014867.1 | 4 - 5 | 449 | 149 | |
GenBank mRNA | BC123458.1 | - | 4 - 5 | 452 | 149 | |
BT021515.1 | - | 4 - 5 | 449 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |